2-Amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile
Supplier: Matrix Scientific
011606-500MG
101800-972EA
204.7
USD
101800-972
2-Amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile
2-Amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile
CAS: 37578-06-6
MDL Number: MFCD00712387
Molecular Formula: C9H11N3S
Molecular Weight: 193,27
Hazard: Irritant
Formula:
C₉H₁₁N₃S Melting Pt: 183…185 °C |
MDL Number:
MFCD00712387 CAS Number: 37578-06-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...