6-Chloro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Supplier: Matrix Scientific
011577-500MG
101800-918EA
273.85
USD
101800-918
6-Chloro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
6-Chloro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
CAS: 53977-19-8
MDL Number: MFCD01829307
Molecular Formula: C10H6ClNO3
Molecular Weight: 223,62
Hazard: Irritant
Formula:
C₁₀H₆ClNO₃ |
MDL Number:
MFCD01829307 CAS Number: 53977-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...