5-Methoxy-1,3-benzothiazol-2-amine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-methoxy-1,3-benzothiazole

011563-1G
101800-890EA 79.35 USD
101800-890
5-Methoxy-1,3-benzothiazol-2-amine ≥98%
5-Methoxy-1,3-benzothiazol-2-amine

Purity: 98%
CAS: 54346-87-1
MDL Number: MFCD00464314
Molecular Formula: C8H8N2OS
Molecular Weight: 180,23
Hazard: Irritant

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Melting Pt: 154 °C
MDL Number: MFCD00464314
CAS Number: 54346-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR