[(4-Methyl-furazan-3-carbonyl)-amino]-acetic acid

Supplier: Matrix Scientific

011545-500MG
101800-856EA 204.7 USD
101800-856
[(4-Methyl-furazan-3-carbonyl)-amino]-acetic acid
[(4-Methylfurazan-3-carbonyl)amino]acetic acid

CAS: 797806-70-3
MDL Number: MFCD01201960
Molecular Formula: C6H7N3O4
Molecular Weight: 185,14
Hazard: Irritant

Formula: C₆H₇N₃O₄
MDL Number: MFCD01201960
CAS Number: 797806-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR