2,1,3-Benzothiadiazole-5-carboxylic acid

Supplier: Matrix Scientific

011524-500MG
101800-814EA 115.38 USD
101800-814
2,1,3-Benzothiadiazole-5-carboxylic acid
2,1,3-Benzothiadiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₇H₄N₂O₂S
Melting Pt: 226 °C
MDL Number: MFCD01647555
CAS Number: 16405-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR