4-(Morpholinosulfonyl)aniline
Supplier: Matrix Scientific
011498-500MG
101800-766EA
273.85
USD
101800-766
4-(Morpholinosulfonyl)aniline
4-(Morpholinosulphonyl)aniline
Hazard: Irritant
Formula:
C₁₀H₁₄N₂O₃S Melting Pt: 217 °C |
MDL Number:
MFCD00277213 CAS Number: 21626-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...