4-(1-Adamantyl)-2-aminothiazole hydrochloride

Supplier: Matrix Scientific

011487-500MG
101800-748EA 204.7 USD
101800-748
4-(1-Adamantyl)-2-aminothiazole hydrochloride
4-(1-Adamantyl)-2-aminothiazole hydrochloride

CAS: 462605-77-2
MDL Number: MFCD01825559
Molecular Formula: C13H19ClN2S
Molecular Weight: 270,83
Hazard: Irritant

Formula: C₁₃H₁₉ClN₂S
MDL Number: MFCD01825559
CAS Number: 462605-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR