3-Amino-4-methylbenzenesulfonamide

Supplier: Matrix Scientific

011428-500MG
101800-630EA 76.92 USD
101800-630
3-Amino-4-methylbenzenesulfonamide
3-Amino-4-methylbenzenesulphonamide

Hazard: Irritant

Formula: C₇H₁₀N₂O₂S
Melting Pt: 176 °C
MDL Number: MFCD00188937
CAS Number: 6274-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR