4-Morpholinobenzaldehyde

Supplier: Matrix Scientific

011418-500MG
101800-610EA 15.38 USD
101800-610
4-Morpholinobenzaldehyde
4-Morpholinobenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MW: 191.2264 g/mol
MDL Number: MFCD00735826
CAS Number: 1204-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR