3-Methyl-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

011355-500MG
101800-486EA 69.23 USD
101800-486
3-Methyl-1H-pyrazole-4-carboxylic acid
3-Methyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₅H₆N₂O₂
MW: 126.1133 g/mol
Melting Pt: 230 °C
MDL Number: MFCD00608153
CAS Number: 40704-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR