2-Hydroxyisonicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-4-pyridinecarboxylic acid

011249-25G 011249-5G
101800-212EA 16.92 USD
101800-212 101800-214
2-Hydroxyisonicotinic acid ≥98%
2-Hydroxyisonicotinic acid

Hazard: Irritant

Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD01318566
CAS Number: 22282-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR