Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Tributylstannyl)benzo[b]thiophene

Supplier: Matrix Scientific

Danger

011161-1G
101800-026EA 122.92 USD InStock
101800-026
2-(Tributylstannyl)benzo[b]thiophene
2-(Tributylstannyl)benzo[b]thiophene
CAS: 148961-88-0
MDL Number: MFCD01318936
Molecular Formula: C20H32SSn
Molecular Weight: 423,23
Hazard: Irritant/Refrigerate


Formula: C₂₀H₃₂SSn
MW: 423,22
MDL Number: MFCD01318936
CAS Number: 148961-88-0
EINECS: 000-000-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty