3-Fluoro-4-nitrobenzonitrile ≥98%

Supplier: Matrix Scientific

011126-5G
101799-942EA 61.54 USD
101799-942
3-Fluoro-4-nitrobenzonitrile ≥98%
3-Fluoro-4-nitrobenzonitrile

Hazard: Irritant

Formula: C₇H₃FN₂O₂
MW: 166.1093 g/mol
Melting Pt: 72…73 °C
MDL Number: MFCD04973417
CAS Number: 218632-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR