2,6-Dichloro-3-iodopyridine ≥98%

Supplier: Matrix Scientific

011122-5G 011122-25G
101799-928EA 72.31 USD
101799-928 101799-930
2,6-Dichloro-3-iodopyridine ≥98%
2,6-Dichloro-3-iodopyridine

Hazard: Irritant

Formula: C₅H₂Cl₂IN
MW: 273.88 g/mol
MDL Number: MFCD03094686
CAS Number: 148493-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR