2,4-Dichloro-5-fluorobenzamide

Supplier: Matrix Scientific

005105-1G 005105-5G
101788-254EA 155.38 USD
101788-254 101788-252
2,4-Dichloro-5-fluorobenzamide
2,4-Dichloro-5-fluorobenzamide

Hazard: Irritant

Formula: C₇H₄Cl₂FNO
Melting Pt: 158…160 °C
MDL Number: MFCD01631478
CAS Number: 135748-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR