(2,3-Difluoro-4-methylphenyl)acetonitrile
Supplier: Matrix Scientific
005080-5G
005080-1G
101788-236EA
103.08
USD
101788-236
101788-238
(2,3-Difluoro-4-methylphenyl)acetonitrile
(2,3-Difluoro-4-methylphenyl)acetonitrile
Hazard: Toxic
Formula:
C₉H₇F₂N |
MDL Number:
MFCD01631648 CAS Number: 261944-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...