(2,3-Difluor-4-methylphenyl)acetic acid

Supplier: Matrix Scientific

005079-1G
101788-234EA 147.69 USD
101788-234
(2,3-Difluor-4-methylphenyl)acetic acid
(2,3-Difluor-4-methylphenyl)acetic acid

Hazard: Irritant

Formula: C₉H₈F₂O₂
MW: 186.16
Melting Pt: 124…126 °C
MDL Number: MFCD01631649
CAS Number: 261763-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR