2,3-Difluoro-4-(trifluoromethyl)benzoic acid

Supplier: Matrix Scientific

005034-1G
101788-190EA 58.46 USD
101788-190
2,3-Difluoro-4-(trifluoromethyl)benzoic acid
2,3-Difluoro-4-(trifluoromethyl)benzoic acid

Hazard: Irritant

Formula: C₈H₃F₅O₂
Melting Pt: 119…122 °C
MDL Number: MFCD00236256
CAS Number: 237424-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR