(Perfluoro-n-octyl)phenyliodonium trifluoromethanesulfonate

Supplier: Matrix Scientific

Synonyms: (Eicosafluoro-n-octyl)phenyliodonium Trifluoromethanesulfonate(Eicosafluoro-n-octyl)phenyliodonium Trifluoromethanesulphonate

004811-500MG
101787-848EA 256.92 USD
101787-848
(Perfluoro-n-octyl)phenyliodonium trifluoromethanesulfonate
(Perfluoro-n-octyl)phenyliodonium trifluoromethanesulfonate

Hazard: Irritant

Formula: C₁₅H₅F₂₀IO₃S
MW: 772.13 g/mol
MDL Number: MFCD00135120
CAS Number: 77758-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR