Methyl pentafluorobenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl Pentafluorobenzoate

004696-100G
101787-688EA 192.05 USD
101787-688
Methyl pentafluorobenzoate ≥97%
Methyl pentafluorobenzoate

Hazard: Irritant

Formula: C₈H₃F₅O₂
MW: 226.1002 g/mol
Boiling Pt: 72 °C (15 Torr)
MDL Number: MFCD00012172
CAS Number: 36629-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR