DBD-F [=4-(N,N-dimethylaminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole]

Supplier: Matrix Scientific

Synonyms: 7-Fluoro-4-(N,N-dimethylaminosulphonyl)benzofurazan4-(N,N-Dimethylaminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole7-Fluoro-4-(N,N-dimethylaminosulfonyl)benzofurazan7-Fluoro-4-(N,N-dimethylsulphamoyl)-2,1,3-benzoxadiazole4-(N,N-Dimethylaminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole7-Fluoro-4-(N,N-dimethylsulfamoyl)-2,1,3-benzoxadiazole

004485-100MG
101787-312EA 283.08 USD
101787-312
DBD-F [=4-(N,N-dimethylaminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole]
DBD-F [=4-(N,N-dimethylaminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole]

CAS: 98358-90-8
MDL Number: MFCD00143356
Molecular Formula: C8H8FN3O3S
Molecular Weight: 245,23
Hazard: Irritant

Formula: C₈H₈FN₃O₃S
MW: 245.23 g/mol
MDL Number: MFCD00143356
CAS Number: 98358-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR