4-Fluorodiphenylmethane ≥99%

Supplier: Matrix Scientific

004221-5G 004221-1G
101786-982EA 27.6 USD
101786-982 101786-984
4-Fluorodiphenylmethane ≥99%
4-Fluorodiphenylmethane

Hazard: Irritant

Formula: C₁₃H₁₁F
Boiling Pt: 255…256 °C (748 Torr)
Density: 1.0909
MDL Number: MFCD00464329
CAS Number: 587-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR