2,4-Dimethoxy-5-pyrimidineboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (2,4-Dimethoxypyrimidin-5-yl)boronic acid2,4-Dimethoxypyrimidine-5-boronic acid2,4-Dimethoxy-5-pyrimidylboronic acid

004024-1G
101786-694EA 27.6 USD
101786-694
2,4-Dimethoxy-5-pyrimidineboronic acid ≥97%
2,4-Dimethoxy-5-pyrimidineboronic acid

Hazard: Irritant/Refrigerate

Formula: C₆H₉BN₂O₄
MW: 183.9577 g/mol
Melting Pt: 113…117 °C
MDL Number: MFCD01318985
CAS Number: 89641-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR