9-Decenoic acid ≥90%

Supplier: Matrix Scientific

003056-5G 003056-25G
101785-144EA 216.92 USD
101785-144 101785-142
9-Decenoic acid ≥90%
9-Decenoic acid

Hazard: Irritant

Formula: C₁₀H₁₈O₂
Boiling Pt: 142 °C (4 Torr)
Melting Pt: <0 °C
MDL Number: MFCD00036663
CAS Number: 14436-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR