2-Amino-4-(2,4-difluorophenyl)thiazole

Supplier: Matrix Scientific

028803-500MG
101826-880EA 193.85 USD
101826-880
2-Amino-4-(2,4-difluorophenyl)thiazole
2-Amino-4-(2,4-difluorophenyl)thiazole

Hazard: Irritant

Formula: C₉H₆F₂N₂S
MW: 212.22
MDL Number: MFCD00472891
CAS Number: 105512-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR