2-Amino-4-(4-nitrophenyl)thiazole

Supplier: Matrix Scientific

028801-500MG
101826-876EA 40 USD
101826-876
2-Amino-4-(4-nitrophenyl)thiazole
2-Amino-4-(4-nitrophenyl)thiazole

Hazard: Irritant

Formula: C₉H₇N₃O₂S
Melting Pt: 285…286 °C
MDL Number: MFCD00170222
CAS Number: 2104-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR