4-Amino-1-ethylpiperidine

Supplier: Matrix Scientific

028774-500MG
101826-822EA 78.46 USD
101826-822
4-Amino-1-ethylpiperidine
4-Amino-1-ethylpiperidine

Hazard: Irritant

Formula: C₇H₁₆N₂
MW: 128.22 g/mol
MDL Number: MFCD03640283
CAS Number: 50534-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR