1-(2,4-Dichlorobenzyl)piperazine

Supplier: Matrix Scientific

028766-500MG
101826-804EA 193.85 USD
101826-804
1-(2,4-Dichlorobenzyl)piperazine
1-(2,4-Dichlorobenzyl)piperazine

Hazard: Irritant

Formula: C₁₁H₁₄Cl₂N₂
MDL Number: MFCD03407487
CAS Number: 51619-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR