Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-α-Ethylbenzylamine

Supplier: Matrix Scientific


028749-500MG
101826-766EA 20.92 USD InStock
101826-766
(±)-α-Ethylbenzylamine
(±)-α-Ethylbenzylamine
CAS: 2941-20-0
MDL Number: MFCD00038169
Molecular Formula: C9H13N
Molecular Weight: 135,21
Hazard: Irritant


Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty