2-[(2,4-Dichlorobenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028709-500MG
101826-686EA 146.15 USD
101826-686
2-[(2,4-Dichlorobenzyl)oxy]benzaldehyde
2-[(2,4-Dichlorobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀Cl₂O₂
MDL Number: MFCD01025492
CAS Number: 52803-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR