3-Ethoxy-4-[(3-fluorobenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028635-500MG
101826-536EA 109.25 USD
101826-536
3-Ethoxy-4-[(3-fluorobenzyl)oxy]benzaldehyde
3-Ethoxy-4-[(3-fluorobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₆H₁₅FO₃
MDL Number: MFCD01990236
CAS Number: 346459-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR