4-[(3-Fluorobenzyl)oxy]-3-methoxybenzaldehyde

Supplier: Matrix Scientific

028623-500MG
101826-512EA 146.15 USD
101826-512
4-[(3-Fluorobenzyl)oxy]-3-methoxybenzaldehyde
4-[(3-Fluorobenzyl)oxy]-3-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₃FO₃
MDL Number: MFCD02255724
CAS Number: 428473-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR