3-Methoxy-4-[(2-methylbenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028619-500MG
101826-504EA 146.15 USD
101826-504
3-Methoxy-4-[(2-methylbenzyl)oxy]benzaldehyde
3-Methoxy-4-[(2-methylbenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₆H₁₆O₃
MDL Number: MFCD02647956
CAS Number: 361465-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR