4-[(4-Methylbenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028613-500MG
101826-492EA 86.15 USD
101826-492
4-[(4-Methylbenzyl)oxy]benzaldehyde
4-[(4-Methylbenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₄O₂
MDL Number: MFCD00617729
CAS Number: 66742-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR