4-[(3-(Trifluoromethyl)benzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028609-500MG
101826-484EA 146.15 USD
101826-484
4-[(3-(Trifluoromethyl)benzyl)oxy]benzaldehyde
4-[(3-(Trifluoromethyl)benzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₁F₃O₂
MDL Number: MFCD03422467
CAS Number: 70627-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR