4-[(2,6-Dichlorobenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028605-500MG
101826-476EA 146.15 USD
101826-476
4-[(2,6-Dichlorobenzyl)oxy]benzaldehyde
4-[(2,6-Dichlorobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀Cl₂O₂
MDL Number: MFCD03422465
CAS Number: 166049-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR