4-Methoxy-3-[(3-methylbenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028594-500MG
101826-450EA 146.15 USD
101826-450
4-Methoxy-3-[(3-methylbenzyl)oxy]benzaldehyde
4-Methoxy-3-[(3-methylbenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₆H₁₆O₃
MDL Number: MFCD03422463
CAS Number: 667412-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR