3-[(2-Chloro-6-fluorobenzyl)oxy]-4-methoxybenzaldehyde

Supplier: Matrix Scientific

028590-500MG
101826-442EA 109.25 USD
101826-442
3-[(2-Chloro-6-fluorobenzyl)oxy]-4-methoxybenzaldehyde
3-[(2-Chloro-6-fluorobenzyl)oxy]-4-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₂ClFO₃
MDL Number: MFCD01590677
CAS Number: 591224-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR