3-[(3-Methoxybenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028582-500MG
101826-426EA 109.25 USD
101826-426
3-[(3-Methoxybenzyl)oxy]benzaldehyde
3-[(3-Methoxybenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MDL Number: MFCD03422457
CAS Number: 588685-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR