3-[(2-Chloro-4-fluorobenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028578-500MG
101826-418EA 64.4 USD
101826-418
3-[(2-Chloro-4-fluorobenzyl)oxy]benzaldehyde
3-[(2-Chloro-4-fluorobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀ClFO₂
MDL Number: MFCD03422453
CAS Number: 588681-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR