3-[(3-Fluorobenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028576-500MG
101826-414EA 146.15 USD
101826-414
3-[(3-Fluorobenzyl)oxy]benzaldehyde
3-[(3-Fluorobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₁FO₂
MDL Number: MFCD03422452
CAS Number: 590353-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR