3-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028574-500MG
101826-410EA 146.15 USD
101826-410
3-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde
3-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀ClFO₂
MDL Number: MFCD01590673
CAS Number: 336879-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR