5-Chloro-2-[(3-methoxybenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028552-500MG
101826-366EA 109.25 USD
101826-366
5-Chloro-2-[(3-methoxybenzyl)oxy]benzaldehyde
5-Chloro-2-[(3-methoxybenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₃ClO₃
MDL Number: MFCD03422436
CAS Number: 667412-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR