5-Chloro-2-[(2-methylbenzyl)oxy]benzaldehyde
Supplier: Matrix Scientific
028550-500MG
101826-362EA
146.15
USD
101826-362
5-Chloro-2-[(2-methylbenzyl)oxy]benzaldehyde
5-Chloro-2-[(2-methylbenzyl)oxy]benzaldehyde
Hazard: Irritant
Formula:
C₁₅H₁₃ClO₂ |
MDL Number:
MFCD03422434 CAS Number: 590360-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...