5-Chloro-2-[(2-chloro-4-fluorobenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028548-500MG
101826-358EA 109.25 USD
101826-358
5-Chloro-2-[(2-chloro-4-fluorobenzyl)oxy]benzaldehyde
5-Chlor-2-[(2-chloro-4-fluorobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₄H₉Cl₂FO₂
MDL Number: MFCD03422432
CAS Number: 667436-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR