5-Chloro-2-[(3-fluorobenzyl)oxy]benzaldehyde
Supplier: Matrix Scientific
028546-500MG
101826-354EA
146.15
USD
101826-354
5-Chloro-2-[(3-fluorobenzyl)oxy]benzaldehyde
5-Chloro-2-[(3-fluorobenzyl)oxy]benzaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₀ClFO₂ |
MDL Number:
MFCD03422430 CAS Number: 667437-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...