5-Chloro-2-[(2-chloro-6-fluorobenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028542-500MG
101826-346EA 109.25 USD
101826-346
5-Chloro-2-[(2-chloro-6-fluorobenzyl)oxy]benzaldehyde
5-Chlor-2-[(2-chloro-6-fluorobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₄H₉Cl₂FO₂
MDL Number: MFCD03422426
CAS Number: 667436-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR