3-Methoxy-2-[(3-methoxybenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

028518-500MG
101826-298EA 146.15 USD
101826-298
3-Methoxy-2-[(3-methoxybenzyl)oxy]benzaldehyde
3-Methoxy-2-[(3-methoxybenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₆H₁₆O₄
MDL Number: MFCD03422412
CAS Number: 667412-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR