3-Methoxy-2-[(2-methylbenzyl)oxy]benzaldehyde
Supplier: Matrix Scientific
028516-500MG
101826-294EA
146.15
USD
101826-294
3-Methoxy-2-[(2-methylbenzyl)oxy]benzaldehyde
3-Methoxy-2-[(2-methylbenzyl)oxy]benzaldehyde
Hazard: Irritant
Formula:
C₁₆H₁₆O₃ |
MDL Number:
MFCD03422410 CAS Number: 588681-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...