2-[(2-Chloro-4-fluorobenzyl)oxy]-3-methoxybenzaldehyde

Supplier: Matrix Scientific

028514-500MG
101826-290EA 146.15 USD
101826-290
2-[(2-Chloro-4-fluorobenzyl)oxy]-3-methoxybenzaldehyde
2-[(2-Chloro-4-fluorobenzyl)oxy]-3-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₂ClFO₃
MDL Number: MFCD03422408
CAS Number: 588692-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR